Learn More
Medchemexpress LLC HY-B0305A 500mg Medchemexpress, Roxatidine (Acetate Hydrochloride) CAS:93793-83-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0305A500MG
Encompass_Preferred
Medchemexpress, HY-B0305A 500mg Roxatidine (Acetate Hydrochloride) CAS:93793-83-0 Roxatidine Acetate Hydrochloride (HOE 760) is a selective histamine H2 receptor antagonist, can be used for the research of gastric and duodenal ulcers. Roxatidine Acetate Hydrochloride can be rapidly converted to its active metabolite, roxatidine, by esterases in the small intestine, plasma, and liver. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2458