Learn More
Medchemexpress LLC Roxatidine Acetate Hydrochloride (HOE 760) | 93793-83-0 | 99.6% | 1 G
SDP
Catalog No. 5000406549
Encompass_Preferred
Roxatidine Acetate Hydrochloride is a potent, selective, competitive, and orally active histamine H2-receptor antagonist. It demonstrates antisecretory potency against gastric acid secretion and can suppress inflammatory responses. It is used for gastric and duodenal ulcers research and also exhibits antitumor activity.
  • Potent, selective, competitive, and orally active histamine H2-receptor antagonist
  • Demonstrates antisecretory potency against gastric acid secretion
  • Suppresses inflammatory responses by inhibiting NF-κB and p38 MAPK activation
  • Used for gastric and duodenal ulcers research
  • Exhibits antitumor activity
  • Suppresses growth of colon 38 tumor implants in mice
Catalog No. 50-004-06549 Supplier Medchemexpress LLC Supplier No. HYB0305A1G
Roxatidine Acetate Hydrochloride is a potent, selective, competitive, and orally active histamine H2-receptor antagonist. It demonstrates antisecretory potency against gastric acid secretion and can suppress inflammatory responses. It is used for gastric and duodenal ulcers research and also exhibits antitumor activity.
  • Potent, selective, competitive, and orally active histamine H2-receptor antagonist
  • Demonstrates antisecretory potency against gastric acid secretion
  • Suppresses inflammatory responses by inhibiting NF-κB and p38 MAPK activation
  • Used for gastric and duodenal ulcers research
  • Exhibits antitumor activity
  • Suppresses growth of colon 38 tumor implants in mice