Learn More
TARGETMOL CHEMICALS INC ROXIFIBAN ACETATE 5MG
SDP
Catalog No. 502580786
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Roxifiban acetate(DMP 754 acetate) is a potent GP IIb / IIIa antagonist that exhibits antiplatelet aggregation activity via immune mediation and can be used in the study of human thrombocytopenia. purity: 99%
Catalog No. 50-258-0786 Supplier TARGETMOL CHEMICALS INC Supplier No. T247335MG
Add to Cart
Add to Cart
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Roxifiban acetate(DMP 754 acetate) is a potent GP IIb / IIIa antagonist that exhibits antiplatelet aggregation activity via immune mediation and can be used in the study of human thrombocytopenia. purity: 99%