Learn More
Medchemexpress LLC Roxindole hydrochloride | 108050-82-4 | 99.0% | 382.93 g/mol | C23H27ClN2O | 1 ML
SDP
Catalog No. 5000243850
Encompass_Preferred
Roxindole hydrochloride is a research chemical acting as a dopamine autoreceptor agonist and a 5-HT1A receptor agonist with reported antipsychotic and antidepressant activity. It is intended for laboratory research and analytical use and is supplied in solution and solid forms with accompanying documentation for traceability and safety.
Catalog No. 50-002-43850 Supplier Medchemexpress LLC Supplier No. HY106100A10MM 1ML
Roxindole hydrochloride is a research chemical acting as a dopamine autoreceptor agonist and a 5-HT1A receptor agonist with reported antipsychotic and antidepressant activity. It is intended for laboratory research and analytical use and is supplied in solution and solid forms with accompanying documentation for traceability and safety.