Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Roxindole hydrochloride | 108050-82-4 | 99.5% | 382.93 g·mol⁻1 | C23H27ClN2O | 50 MG
SDP

Catalog No. 5000243528
Encompass_Preferred

Roxindole hydrochloride is a research-grade small molecule used as a pharmacological tool to study serotonin and dopamine receptor activity. It functions as a 5-HT1A agonist and dopaminergic modulator and is provided as a hydrochloride salt for use in in vitro and biochemical assays.

  • Used to probe 5-HT1A and dopamine receptor pharmacology
  • High purity suitable for research assays (HPLC)
  • Molecular weight 382.93 g·mol⁻1
  • Chemical formula C23H27ClN2O; CAS 108050-82-4
  • Available in small solid quantities for laboratory experiments
  • Store solid at -20°C and protect from light

Catalog No. 50-002-43528 Supplier Medchemexpress LLC Supplier No. HY106100A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Roxindole hydrochloride is a research-grade small molecule used as a pharmacological tool to study serotonin and dopamine receptor activity. It functions as a 5-HT1A agonist and dopaminergic modulator and is provided as a hydrochloride salt for use in in vitro and biochemical assays.

  • Used to probe 5-HT1A and dopamine receptor pharmacology
  • High purity suitable for research assays (HPLC)
  • Molecular weight 382.93 g·mol⁻1
  • Chemical formula C23H27ClN2O; CAS 108050-82-4
  • Available in small solid quantities for laboratory experiments
  • Store solid at -20°C and protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.