Learn More
Selleck Chemical LLC Roxithromycin S2506-50mg
SDP
Catalog No. 503111633
Encompass_Preferred
Roxithromycin (Roxl-150 Roxo Surlid Rulide Biaxsig Roxar Roximycin Roxomycin Tirabicin Coroxin RU-28965) is a semi-synthetic macrolide antibiotic It is used to treat respiratory tract urinary and soft tissue infections
Catalog No. 50-311-1633 Supplier Selleck Chemical LLC Supplier No. S250650MG
Roxithromycin (Roxl-150 Roxo Surlid Rulide Biaxsig Roxar Roximycin Roxomycin Tirabicin Coroxin RU-28965) is a semi-synthetic macrolide antibiotic It is used to treat respiratory tract urinary and soft tissue infections