Learn More
Medchemexpress LLC Roxithromycin | 80214-83-1 | 1 G
SDP
Catalog No. 5000409305
Encompass_Preferred
Roxithromycin (RU-28965) is an orally active semi-synthetic macrolide antibiotic. It inhibits protein biosynthesis in the elongation step by binding to the 50S bacterial ribosome. Roxithromycin exhibits antimicrobial, antiproliferative, anti-inflammatory, tumor vasculature inhibiting, and lung injury ameliorating effects.
  • Inhibits protein biosynthesis by binding to 50S bacterial ribosome
  • Active against Gram-positive and Gram-negative bacteria
  • Displays antimicrobial, antiproliferative, and anti-inflammatory effects
  • Inhibits tumor vasculature and ameliorates lung injury
  • Reduces UVB-induced oxidative stress and keratinocyte apoptosis
  • Induces apoptosis in senescent cells
  • Protects against various bacterial infections in vivo
  • Inhibits tumor angiogenesis and attenuates lung injury in animal models
  • Modulates NF-κB and MAPK/ERK pathways
Catalog No. 50-004-09305 Supplier Medchemexpress LLC Supplier No. HYB04351G
Roxithromycin (RU-28965) is an orally active semi-synthetic macrolide antibiotic. It inhibits protein biosynthesis in the elongation step by binding to the 50S bacterial ribosome. Roxithromycin exhibits antimicrobial, antiproliferative, anti-inflammatory, tumor vasculature inhibiting, and lung injury ameliorating effects.
  • Inhibits protein biosynthesis by binding to 50S bacterial ribosome
  • Active against Gram-positive and Gram-negative bacteria
  • Displays antimicrobial, antiproliferative, and anti-inflammatory effects
  • Inhibits tumor vasculature and ameliorates lung injury
  • Reduces UVB-induced oxidative stress and keratinocyte apoptosis
  • Induces apoptosis in senescent cells
  • Protects against various bacterial infections in vivo
  • Inhibits tumor angiogenesis and attenuates lung injury in animal models
  • Modulates NF-κB and MAPK/ERK pathways