Learn More
Apexbio Technology LLC Roxithromycin 80214-83-1 1g
SDP
Catalog No. 5000092884
Encompass_Preferred
Roxithromycin (80214-83-1) is a semisynthetic macrolide antibiotic that acts by binding to the bacterial ribosomal 50S subunit thereby inhibiting peptide chain elongation and disrupting protein synthesis Roxithromycin exerts its biological activity primarily through this mechanism of action In both in vitro and in vivo models Roxithromycin demonstrates inhibitory effects on cell proliferation inflammation pathways and tumor angiogenesis Reported IC50 values typically vary depending on the cell type and assay methodology Based on these pharmacological properties Roxithromycin holds research potential in studies of inflammation-associated pathologies cancer biology and lung injury
Catalog No. 50-000-92884 Supplier Apexbio Technology LLC Supplier No. B20241000
Roxithromycin (80214-83-1) is a semisynthetic macrolide antibiotic that acts by binding to the bacterial ribosomal 50S subunit thereby inhibiting peptide chain elongation and disrupting protein synthesis Roxithromycin exerts its biological activity primarily through this mechanism of action In both in vitro and in vivo models Roxithromycin demonstrates inhibitory effects on cell proliferation inflammation pathways and tumor angiogenesis Reported IC50 values typically vary depending on the cell type and assay methodology Based on these pharmacological properties Roxithromycin holds research potential in studies of inflammation-associated pathologies cancer biology and lung injury