Learn More
Medchemexpress LLC Roxithromycin | 80214-83-1 | 5 G
SDP
Catalog No. 5000407896
Encompass_Preferred
Roxithromycin is an orally active semi-synthetic macrolide antibiotic that inhibits protein biosynthesis by binding to the 50S bacterial ribosome during the elongation step. It demonstrates antimicrobial, antiproliferative, anti-inflammatory, and tumor vasculature inhibiting effects, also ameliorating lung injury.
  • Orally active semi-synthetic macrolide antibiotic
  • Inhibits protein biosynthesis in bacterial ribosomes
  • Effective against Gram-positive and Gram-negative bacteria
  • Exhibits antiproliferative and anti-inflammatory effects
  • Inhibits endothelial cell migration and tube formation
  • Reduces UV B irradiation-induced reactive oxygen intermediates
  • Induces apoptosis to eliminate senescent cells
  • Attenuates bleomycin-induced lung injury and fibrosis
  • Inhibits constitutive activation of NF-κB and attenuates airway inflammation
Catalog No. 50-004-07896 Supplier Medchemexpress LLC Supplier No. HYB04355G
Roxithromycin is an orally active semi-synthetic macrolide antibiotic that inhibits protein biosynthesis by binding to the 50S bacterial ribosome during the elongation step. It demonstrates antimicrobial, antiproliferative, anti-inflammatory, and tumor vasculature inhibiting effects, also ameliorating lung injury.
  • Orally active semi-synthetic macrolide antibiotic
  • Inhibits protein biosynthesis in bacterial ribosomes
  • Effective against Gram-positive and Gram-negative bacteria
  • Exhibits antiproliferative and anti-inflammatory effects
  • Inhibits endothelial cell migration and tube formation
  • Reduces UV B irradiation-induced reactive oxygen intermediates
  • Induces apoptosis to eliminate senescent cells
  • Attenuates bleomycin-induced lung injury and fibrosis
  • Inhibits constitutive activation of NF-κB and attenuates airway inflammation