Learn More
Medchemexpress LLC Roxithromycin (Standard) | 80214-83-1 | 250 MG
SDP
Catalog No. 5000406140
Encompass_Preferred
Roxithromycin (Standard) is an analytical standard of Roxithromycin, a semi-synthetic macrolide antibiotic (RU-28965) intended for research and analytical applications. It inhibits protein biosynthesis by binding to the 50S bacterial ribosome and exhibits antimicrobial, antiproliferative, anti-inflammatory, tumour vasculature inhibiting, and lung injury ameliorating effects.
  • Intended for research and analytical applications
  • Functions as an analytical standard
  • Inhibits protein biosynthesis by binding to 50S bacterial ribosome
  • Demonstrates antimicrobial effects
  • Exhibits antiproliferative and anti-inflammatory properties
  • Shows tumor vasculature inhibiting and lung injury ameliorating effects
Catalog No. 50-004-06140 Supplier Medchemexpress LLC Supplier No. 250MGHYB0435R
Roxithromycin (Standard) is an analytical standard of Roxithromycin, a semi-synthetic macrolide antibiotic (RU-28965) intended for research and analytical applications. It inhibits protein biosynthesis by binding to the 50S bacterial ribosome and exhibits antimicrobial, antiproliferative, anti-inflammatory, tumour vasculature inhibiting, and lung injury ameliorating effects.
  • Intended for research and analytical applications
  • Functions as an analytical standard
  • Inhibits protein biosynthesis by binding to 50S bacterial ribosome
  • Demonstrates antimicrobial effects
  • Exhibits antiproliferative and anti-inflammatory properties
  • Shows tumor vasculature inhibiting and lung injury ameliorating effects