Learn More
Medchemexpress LLC Roxithromycin Standard | 80214-83-1 | 98.2% | 100 MG
SDP
Catalog No. 5000408602
Encompass_Preferred
Roxithromycin (Standard) is an analytical standard and a semi-synthetic macrolide antibiotic. It is intended for research and analytical applications, inhibiting protein biosynthesis by binding to the 50S bacterial ribosome. Roxithromycin exhibits antimicrobial, antiproliferative, anti-inflammatory, tumour vasculature inhibiting, and lung injury ameliorating effects.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Inhibits protein biosynthesis in the elongation step.
  • Possesses antimicrobial effects.
  • Exhibits antiproliferative effects.
  • Acts as an anti-inflammatory agent.
  • Inhibits tumour vasculature.
  • Ameliorates lung injury.
Catalog No. 50-004-08602 Supplier Medchemexpress LLC Supplier No. HYB0435R100MG
Roxithromycin (Standard) is an analytical standard and a semi-synthetic macrolide antibiotic. It is intended for research and analytical applications, inhibiting protein biosynthesis by binding to the 50S bacterial ribosome. Roxithromycin exhibits antimicrobial, antiproliferative, anti-inflammatory, tumour vasculature inhibiting, and lung injury ameliorating effects.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Inhibits protein biosynthesis in the elongation step.
  • Possesses antimicrobial effects.
  • Exhibits antiproliferative effects.
  • Acts as an anti-inflammatory agent.
  • Inhibits tumour vasculature.
  • Ameliorates lung injury.