Learn More
Medchemexpress LLC Roxithromycin (Standard) | 80214-83-1 | 98.2% | 50 MG
SDP
Catalog No. 5000405303
Encompass_Preferred
Roxithromycin (Standard), also known as RU-28965 (Standard), is an analytical standard for Roxithromycin. This product is designed for research and analytical applications. Roxithromycin (RU-28965) itself is a semi-synthetic macrolide antibiotic. As an analytical standard, it serves as a reference standard for assays and is frequently utilized in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Analytical standard for roxithromycin
  • Intended for research and analytical applications
  • Serves as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-004-05303 Supplier Medchemexpress LLC Supplier No. HYB0435R50MG
Roxithromycin (Standard), also known as RU-28965 (Standard), is an analytical standard for Roxithromycin. This product is designed for research and analytical applications. Roxithromycin (RU-28965) itself is a semi-synthetic macrolide antibiotic. As an analytical standard, it serves as a reference standard for assays and is frequently utilized in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Analytical standard for roxithromycin
  • Intended for research and analytical applications
  • Serves as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS