Learn More
Surgical Design Royaltek™ Carbon Steel Scalpel Blades
Catalog No. 22079775 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 24
# 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-079-775 # 25
22-079-774 # 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-079-775 Supplier Surgical Design Supplier No. BLADE #25
Add to Cart
Add to Cart
Surgical Design™ Royaltek™ Carbon Steel Scalpel Blades are made from high quality carbon steel and sharpened to perfection. 10 series blades fit 3, 3L, 3 Graduated, 5B, 7, 9 B3, and B3L scalpel handles. 20 series blades fit 4, 4L, 4 Graduated and 6B.
Surgical Design™ presents Royaltek™- our premium line of surgical blades
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical grade, carbon-steel blades
  • Individually sealed
  • Peelable packing
  • Sterile
Specifications
Blade Material Carbon Steel
Product Type Scalpel Blade
For Use With (Application) Used for Making Long Incisions in General Surgery and Autopsy Procedures
Size # 25
For Use With (Equipment) Fits # 4, # 4L, # 4 Graduated and # 6B Handles
Description Horizontal Straight Edge Bent at an Angle
Sterility Sterile