Learn More
Selleck Chemical LLC RP-102124 E4622-1G
SDP
Catalog No. 5000038937
Encompass_Preferred
RP-102124 is a first-in-class allosteric inhibitor of multi-Rac with activity against oncogenic Rac1b RP-102124 is orally bioavailable and exhibits dose-dependent inhibition of tumor growth in xenograft models It exhibits potent inhibition of phenotypic characteristics of Rac signaling such as cell migration and cytoskeletal rearrangements
Catalog No. 50-000-38937 Supplier Selleck Chemical LLC Supplier No. E46221G
RP-102124 is a first-in-class allosteric inhibitor of multi-Rac with activity against oncogenic Rac1b RP-102124 is orally bioavailable and exhibits dose-dependent inhibition of tumor growth in xenograft models It exhibits potent inhibition of phenotypic characteristics of Rac signaling such as cell migration and cytoskeletal rearrangements