Learn More
Miele, Inc. RP1150
Catalog No. 501953756
Encompass_Preferred
Roller plinth, 1150 mm. For 1150 mm units with service access from infeed side and installation against a wall. A roller plinth allows the machine to be pulled forward for servicing. Available as an option for PLW 8617.
Catalog No. 50-195-3756 Supplier Miele, Inc. Supplier No. 10941950
Roller plinth, 1150 mm. For 1150 mm units with service access from infeed side and installation against a wall. A roller plinth allows the machine to be pulled forward for servicing. Available as an option for PLW 8617.