Learn More
Medchemexpress LLC RP-1664 | 2980682-00-4 | 98.77% | 514.55 | 5 MG
SDP
Catalog No. 5000385621
Encompass_Preferred
RP-1664 is a selective and orally active PLK4 inhibitor with an IC50 of 3 nM. It demonstrates selectivity over related kinases like AURKA/B and PLK1. This compound disrupts centriole biogenesis in cancer cells, leading to an accumulation of PLK4 and p21 protein. RP-1664 shows increased sensitivity in TRIM37-high-expressing cells or tumors and exhibits anti-tumor activity in breast cancer and neuroblastoma research.
Catalog No. 50-003-85621 Supplier Medchemexpress LLC Supplier No. HY1727375MG
RP-1664 is a selective and orally active PLK4 inhibitor with an IC50 of 3 nM. It demonstrates selectivity over related kinases like AURKA/B and PLK1. This compound disrupts centriole biogenesis in cancer cells, leading to an accumulation of PLK4 and p21 protein. RP-1664 shows increased sensitivity in TRIM37-high-expressing cells or tumors and exhibits anti-tumor activity in breast cancer and neuroblastoma research.