Learn More
Medchemexpress LLC RP-1664 | 2980682-00-4 | 98.77% | 514.55 | 50 MG
SDP
Catalog No. 5000385899
Encompass_Preferred
RP-1664 is a selective and orally active PLK4 inhibitor with an IC50 of 3 nM. It demonstrates exquisite selectivity over related kinases, including AURKA/B and PLK1. It disrupts centriole biogenesis in cancer cells, leading to an accumulation of PLK4 and p21 protein. It shows increased sensitivity in TRIM37-high-expressing cells or tumors and exhibits anti-tumor activity in breast cancer and neuroblastoma research.
  • Selective and orally active PLK4 inhibitor.
  • IC50 of 3 nM for PLK4.
  • Exquisite selectivity over AURKA/B and PLK1.
  • Disrupts centriole biogenesis in cancer cells.
  • Leads to accumulation of PLK4 and p21 protein.
  • Increased sensitivity in TRIM37-high-expressing cells or tumors.
  • Exhibits anti-tumor activity in breast cancer and neuroblastoma research.
Catalog No. 50-003-85899 Supplier Medchemexpress LLC Supplier No. HY17273750MG
RP-1664 is a selective and orally active PLK4 inhibitor with an IC50 of 3 nM. It demonstrates exquisite selectivity over related kinases, including AURKA/B and PLK1. It disrupts centriole biogenesis in cancer cells, leading to an accumulation of PLK4 and p21 protein. It shows increased sensitivity in TRIM37-high-expressing cells or tumors and exhibits anti-tumor activity in breast cancer and neuroblastoma research.
  • Selective and orally active PLK4 inhibitor.
  • IC50 of 3 nM for PLK4.
  • Exquisite selectivity over AURKA/B and PLK1.
  • Disrupts centriole biogenesis in cancer cells.
  • Leads to accumulation of PLK4 and p21 protein.
  • Increased sensitivity in TRIM37-high-expressing cells or tumors.
  • Exhibits anti-tumor activity in breast cancer and neuroblastoma research.