Learn More
Medchemexpress LLC RP-1664 | 2980682-00-4 | 98.8% | 514.55 | 25 MG
SDP
Catalog No. 5000384789
Encompass_Preferred
RP-1664 is a selective and orally active PLK4 inhibitor with an IC50 of 3 nM. It demonstrates high selectivity over related kinases like AURKA/B and PLK1. This compound disrupts centriole biogenesis in cancer cells, leading to an accumulation of PLK4 and p21 protein. RP-1664 shows increased sensitivity in TRIM37-high-expressing cells or tumors and exhibits anti-tumor activity in breast cancer and neuroblastoma research.
  • Selective and orally active PLK4 inhibitor.
  • IC50 of 3 nM for PLK4.
  • Exhibits exquisite selectivity over related kinases such as AURKA/B and PLK1.
  • Disrupts centriole biogenesis in cancer cells.
  • Leads to accumulation of PLK4 and p21 protein.
  • Demonstrates increased sensitivity in TRIM37-high-expressing cells or tumors.
  • Exhibits anti-tumor activity in breast cancer and neuroblastoma research.
  • Appearance: Solid, White to off-white.
Catalog No. 50-003-84789 Supplier Medchemexpress LLC Supplier No. 25MGHY172737
RP-1664 is a selective and orally active PLK4 inhibitor with an IC50 of 3 nM. It demonstrates high selectivity over related kinases like AURKA/B and PLK1. This compound disrupts centriole biogenesis in cancer cells, leading to an accumulation of PLK4 and p21 protein. RP-1664 shows increased sensitivity in TRIM37-high-expressing cells or tumors and exhibits anti-tumor activity in breast cancer and neuroblastoma research.
  • Selective and orally active PLK4 inhibitor.
  • IC50 of 3 nM for PLK4.
  • Exhibits exquisite selectivity over related kinases such as AURKA/B and PLK1.
  • Disrupts centriole biogenesis in cancer cells.
  • Leads to accumulation of PLK4 and p21 protein.
  • Demonstrates increased sensitivity in TRIM37-high-expressing cells or tumors.
  • Exhibits anti-tumor activity in breast cancer and neuroblastoma research.
  • Appearance: Solid, White to off-white.