Learn More
Medchemexpress LLC Rp-1664 | 2980682-00-4 | 98.8% | 514.55 g/mol | C23H24F2N8O2S | 10 MG
SDP
Catalog No. 5000295045
Encompass_Preferred
RP-1664 is a selective, orally active inhibitor of Polo-like kinase 4 (PLK4) used in preclinical oncology and cell-biology research. It potently inhibits PLK4 (IC50 = 3 nM), disrupts centriole biogenesis, and induces centrosome loss in cellular models.
  • Selective PLK4 inhibition (IC50 = 3 nM).
  • Disrupts centriole biogenesis and induces centrosome loss.
  • High purity (98.8%).
  • Molecular weight 514.55 g/mol; formula C23H24F2N8O2S.
  • DMSO solubility 100 mg/mL (ultrasonic recommended).
  • Powder storage -20°C (3 years); in solvent -80°C (6 months).
Catalog No. 50-002-95045 Supplier Medchemexpress LLC Supplier No. HY17273710MG
RP-1664 is a selective, orally active inhibitor of Polo-like kinase 4 (PLK4) used in preclinical oncology and cell-biology research. It potently inhibits PLK4 (IC50 = 3 nM), disrupts centriole biogenesis, and induces centrosome loss in cellular models.
  • Selective PLK4 inhibition (IC50 = 3 nM).
  • Disrupts centriole biogenesis and induces centrosome loss.
  • High purity (98.8%).
  • Molecular weight 514.55 g/mol; formula C23H24F2N8O2S.
  • DMSO solubility 100 mg/mL (ultrasonic recommended).
  • Powder storage -20°C (3 years); in solvent -80°C (6 months).