Learn More
Miles Scientific RP-2 Dimethyl Silanized Silica Gel 60 F on Glass 250U 20X20CM (25 PLATES)
SDP
Catalog No. NC1434762
Encompass_Preferred
RP-2 dimethyl-modified Silica Gel 60 - fluorescent indicator (UV 254) - Glass-backed - 250 µm thickness - 20x20 cm - 25 plates per box This TLC plate can be developed using purely organic, organic/aqueous, and purely aqueous mobile phases. Due to this these plates have been used in numerous applications in normal phase chromatography as well as reversed phase chromatography. Common applications include the analysis of active plant constituents and steroids.
Catalog No. NC1434762 Supplier Miles Scientific Supplier No. P108011
RP-2 dimethyl-modified Silica Gel 60 - fluorescent indicator (UV 254) - Glass-backed - 250 µm thickness - 20x20 cm - 25 plates per box This TLC plate can be developed using purely organic, organic/aqueous, and purely aqueous mobile phases. Due to this these plates have been used in numerous applications in normal phase chromatography as well as reversed phase chromatography. Common applications include the analysis of active plant constituents and steroids.