Learn More
Medchemexpress LLC HY-145817A 5mg , RP-6306 CAS:2719793-90-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY145817A5MG
Encompass_Preferred
HY-145817A 5mg RP-6306 CAS: 2719793-90-3 RP-6306 is a PKMYT1 inhibitor extracted from patent WO2021195781A1 compound 182. RP-6306 can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9191