Learn More
Medchemexpress LLC RP-64477 | 135239-65-5 | 99.6% | 498.72 | 100 MG
SDP
Catalog No. 5000394459
Encompass_Preferred
RP-64477 is a potent inhibitor of the cholesterol esterifying enzyme Acyl-coenzyme A:cholesterol O-acyltransferase (ACAT). It is suitable for various research applications studying cholesterol metabolism.
  • Inhibits ACAT enzyme activity
  • Reduces plasma cholesterol levels in animal models
  • Provided for research use only
Catalog No. 50-003-94459 Supplier Medchemexpress LLC Supplier No. HY16437100MG
RP-64477 is a potent inhibitor of the cholesterol esterifying enzyme Acyl-coenzyme A:cholesterol O-acyltransferase (ACAT). It is suitable for various research applications studying cholesterol metabolism.
  • Inhibits ACAT enzyme activity
  • Reduces plasma cholesterol levels in animal models
  • Provided for research use only