Learn More
Medchemexpress LLC RP-64477 | 135239-65-5 | 99.6% | 498.72 | 5 MG
SDP
Catalog No. 5000394642
Encompass_Preferred
RP-64477 is a potent inhibitor of the cholesterol esterifying enzyme Acyl-coenzyme A:cholesterol O-acyltransferase (ACAT). It significantly reduces plasma cholesterol levels in animal models without affecting food consumption.
  • Potent ACAT inhibitor
  • Inhibits ACAT in various tissue preparations and human cell cultures
  • Reduces plasma cholesterol levels in cholesterol/cholic acid-fed rats
  • No inhibitory activity against rat PCEH or LCAT
Catalog No. 50-003-94642 Supplier Medchemexpress LLC Supplier No. HY164375MG
RP-64477 is a potent inhibitor of the cholesterol esterifying enzyme Acyl-coenzyme A:cholesterol O-acyltransferase (ACAT). It significantly reduces plasma cholesterol levels in animal models without affecting food consumption.
  • Potent ACAT inhibitor
  • Inhibits ACAT in various tissue preparations and human cell cultures
  • Reduces plasma cholesterol levels in cholesterol/cholic acid-fed rats
  • No inhibitory activity against rat PCEH or LCAT