Learn More
Medchemexpress LLC Rp-8-Br-cAMPS sodium | 925456-59-3 | 446.15 | 1 MG
SDP
Catalog No. 5000349197
Encompass_Preferred
Rp-8-Br-cAMPS sodium is an analog of cAMP and an inhibitor of PKA. It occupies cAMP binding sites on PKA type I regulatory subunits, preventing PKA dissociation and activation. It can be used in the study of tumors and retrovirus-induced immune deficiency. Rp-8-Br-cAMPS sodium also inhibits insulin secretion.
  • Completely blocks the ability of 2-chloroadenosine to inhibit LAK cytotoxicity.
  • Significantly reduces the inhibitory effect of 2-chloroadenosine on cytokine production.
  • Inhibits insulin secretion in 832/13 cells.
  • Improves immune function in a mouse retrovirus infection model.
Catalog No. 50-003-49197 Supplier Medchemexpress LLC Supplier No. HY137640C1MG
Add to Cart
Add to Cart
Rp-8-Br-cAMPS sodium is an analog of cAMP and an inhibitor of PKA. It occupies cAMP binding sites on PKA type I regulatory subunits, preventing PKA dissociation and activation. It can be used in the study of tumors and retrovirus-induced immune deficiency. Rp-8-Br-cAMPS sodium also inhibits insulin secretion.
  • Completely blocks the ability of 2-chloroadenosine to inhibit LAK cytotoxicity.
  • Significantly reduces the inhibitory effect of 2-chloroadenosine on cytokine production.
  • Inhibits insulin secretion in 832/13 cells.
  • Improves immune function in a mouse retrovirus infection model.