Learn More
Medchemexpress LLC Rp-8-Br-cAMPS sodium | 925456-59-3 | 446.15 | 500 UG
SDP
Catalog No. 5000351046
Encompass_Preferred
Rp-8-Br-cAMPS sodium is an analog of cAMP and an inhibitor of PKA. It occupies cAMP binding sites on PKA type I regulatory subunits, which prevents PKA dissociation and activation. It can be used in the study of tumors and retrovirus-induced immune deficiency, and it also inhibits insulin secretion.
  • Blocks the ability of 2-chloroadenosine to inhibit LAK cytotoxicity.
  • Significantly reduces the inhibitory effect of 2-chloroadenosine on cytokine production.
  • Inhibits insulin secretion in 832/13 cells.
  • Improves immune function in a mouse retrovirus infection model.
  • Strongly increases responses to the anti-CD3 mAb.
  • Improved T cell responses.
Catalog No. 50-003-51046 Supplier Medchemexpress LLC Supplier No. HY137640C500UG
Rp-8-Br-cAMPS sodium is an analog of cAMP and an inhibitor of PKA. It occupies cAMP binding sites on PKA type I regulatory subunits, which prevents PKA dissociation and activation. It can be used in the study of tumors and retrovirus-induced immune deficiency, and it also inhibits insulin secretion.
  • Blocks the ability of 2-chloroadenosine to inhibit LAK cytotoxicity.
  • Significantly reduces the inhibitory effect of 2-chloroadenosine on cytokine production.
  • Inhibits insulin secretion in 832/13 cells.
  • Improves immune function in a mouse retrovirus infection model.
  • Strongly increases responses to the anti-CD3 mAb.
  • Improved T cell responses.