Learn More
Enzo Life Sciences Rp-8-Br-cGMPS (500µg). CAS: 150418-07-8
SDP
Supplier:  Enzo Life Sciences BMLCN2160500
Encompass_Preferred
Cell-permeable, phosphodiesterase-resistant cGMP antagonist. Reversed NO stimulation of random migration in rabbit neutrophils. Used at 30µM to cometively inhibit 8-Br-cGMP relaxation of rabbit aortas. The product is not sterile. Resistant to nucleotide-dependent phosphodiesterases; no metabolic side effects. Significantly more lipophilic and membrane permeant as compared to cyclic GMP or Rp-cGMPS. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0455