Learn More
Cayman Chemical Rp-8-CPT-CyclIc AMPsodIM500ug
SDP
Catalog No. 502968327
Encompass_Preferred
A lipophilic and non-hydrolyzable cAMP analog that acts as a site-selective inhibitor of PKA type I and II, with preference towards site A of type I and site B of type II
Catalog No. 50-296-8327 Supplier Cayman Chemical Supplier No. 17142500 UG
A lipophilic and non-hydrolyzable cAMP analog that acts as a site-selective inhibitor of PKA type I and II, with preference towards site A of type I and site B of type II