Learn More
Enzo Life Sciences 8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer . triethylammonium salt (100µg). CAS: 153660-04-9
SDP
Supplier:  Enzo Life Sciences ALX480032C100
Encompass_Preferred
Potent selective inhibitor of cGMP-dependent protein kinase 1α, 1β and type II. Resistant to mammalian cyclic nucleotide-dependent phosphodiesterases, no metabolic side effects. Significantly more lipophilic and membrane permeant as compared to Rp-cGMPS (ALX-480-072) or Rp-8-Br-cGMPS (BML-CN216). Alternative name: Rp-8-pCPT-cGMPS . TEA, 8-pCPT-cGMPS . TEA,Rp-Isomer. Purity: ≥99% (HPLC). Appearance: White to off-white crystalline powder. Solubility: Soluble in water and aqueous buffers. Long Term Storage: -20°C.
Catalog No. 50-200-9311