Learn More
Enzo Life Sciences Rp-8-pCPT-cGMPS. sodium salt (1mg). CAS: 153660-04-9
SDP
Catalog No. 502010450
Encompass_Preferred
A cell-permeable cGMP antagonist which is resistant to hydrolysis by cyclic nucleotide phosphodiesterases (Ki=0.5 µM). No metabolic side effects. Significantly more lipophilic and membrane permeant as compared to Rp-cGMPS (ALX-480-072) or Rp-8-Br-cGMPS (BML-CN216) Purity: ≥98% (HPLC). Solubility: Soluble in water (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0450 Supplier Enzo Life Sciences Supplier No. BMLCN2060001
A cell-permeable cGMP antagonist which is resistant to hydrolysis by cyclic nucleotide phosphodiesterases (Ki=0.5 µM). No metabolic side effects. Significantly more lipophilic and membrane permeant as compared to Rp-cGMPS (ALX-480-072) or Rp-8-Br-cGMPS (BML-CN216) Purity: ≥98% (HPLC). Solubility: Soluble in water (25 mg/ml). Long Term Storage: -20°C.