Learn More
Enzo Life Sciences 8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Rp-Isomer . sodium salt (0.5mg). CAS: 129735-00-8
SDP
Supplier:  Enzo Life Sciences ALX480024MC05
Encompass_Preferred
Inhibitor of cAMP-dependent protein kinase (PKA), preferentially binding to type I PKA. Lipophilic analog of Rp-cAMPS (BML-CN135, and ALX-480-085). Resistant to mammalian cyclic nucleotide-dependent phosphodiesterases. Significantly more lipophilic and membrane permeant as compared to Rp-cAMPS or 8-Br-cAMP (BML-CN115). Alternative name: Rp-8-Br-cAMPS . Na, 8-Br-cAMPS . Na, Rp-isomer. Purity: ≥99% (HPLC). Solubility: 250 mM soluble in water, aqueous solutions, DMSO or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-200-9307