Learn More
Miele, Inc. RP900
Catalog No. 501953755
Encompass_Preferred
Roller plinth, 900 mm. For 900 mm units with service access from infeed side and installation against a wall. A roller plinth allows the machine to be pulled forward for servicing. Available as an option for PLW 8615.
Catalog No. 50-195-3755 Supplier Miele, Inc. Supplier No. 10941940
Roller plinth, 900 mm. For 900 mm units with service access from infeed side and installation against a wall. A roller plinth allows the machine to be pulled forward for servicing. Available as an option for PLW 8615.