Learn More
Bon Opus Biosciences Recombinant Human ATP synthase subunit O, mitochondrial/ATP5PO(C-His) - Source: E. coli
SDP
Catalog No. NC2733775
Encompass_Preferred
Recombinant Human ATP synthase subunit O, mitochondrial/ATP5PO is produced by E.coli expression system and the target gene encoding F24-V213 is expressed with a His tag at the C-terminus. Accession#P48047. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733775 Supplier Bon Opus Biosciences Supplier No. BP304350UG
Recombinant Human ATP synthase subunit O, mitochondrial/ATP5PO is produced by E.coli expression system and the target gene encoding F24-V213 is expressed with a His tag at the C-terminus. Accession#P48047. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4