Learn More
Enzo Life Sciences Rp-cAMPS, (1mg), CAS Number: 151837-09-1
SDP
Supplier:  Enzo Life Sciences BMLCN1350001
Encompass_Preferred
PKA inhibitor. Rp-cAMPS is a potent and specific competitive inhibitor of the activation of cAMP-dependent protein kinases by cAMP (IC50=4.9 µM) . Its cell permeability and complete resistance to cyclic nucleotide phosphodiesterases make it a unique tool for studying cAMP-dependent signaling. Significantly more lipophilc as compared to cAMP and thus membrane permeable in many biological systems. This product is not sterile. Formula: C10H11N5O5PS. C6H16N. MW: 446.5. CAS Number: 151837-09-1. Purity: ≥98% (HPLC). Appearance: Clear film. Solubility: Soluble in water (25mg/ml), aqueous buffers, methanol, 100% ethanol or DMSO. Use/Stability: Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C
Catalog No. NC9814297