Learn More
Medchemexpress LLC Rp-cAMPS sodium salt | 142439-94-9 | 99.7% | 367.25 | C10H11N5NaO5PS | 1 MG
SDP
Catalog No. 5000214569
Encompass_Preferred
Rp-cAMPS sodium salt is a cyclic AMP (cAMP) analog that functions as a potent, competitive antagonist of cAMP-dependent protein kinase A (PKA) I and II. Supplied as the sodium salt for research use, it is a white-to-pink solid with high purity and is used to inhibit cAMP-mediated PKA activation in biochemical and cell signaling studies.
  • Potent, competitive antagonist of cAMP-dependent PKA I and II.
  • High purity (99.7%) suitable for research applications.
  • Available in small solid quantities for precise dosing.
  • Stable under recommended storage when sealed and protected from moisture and light.
  • Compatible with biochemical and cell signaling assays to study cAMP pathways.
Catalog No. 50-002-14569 Supplier Medchemexpress LLC Supplier No. HY100530D1MG
Rp-cAMPS sodium salt is a cyclic AMP (cAMP) analog that functions as a potent, competitive antagonist of cAMP-dependent protein kinase A (PKA) I and II. Supplied as the sodium salt for research use, it is a white-to-pink solid with high purity and is used to inhibit cAMP-mediated PKA activation in biochemical and cell signaling studies.
  • Potent, competitive antagonist of cAMP-dependent PKA I and II.
  • High purity (99.7%) suitable for research applications.
  • Available in small solid quantities for precise dosing.
  • Stable under recommended storage when sealed and protected from moisture and light.
  • Compatible with biochemical and cell signaling assays to study cAMP pathways.