Learn More
Medchemexpress LLC Rp-cAMPS sodium salt | 142439-94-9 | 99.7% | 367.25 | C10H11N5NaO5PS | 10 MG
SDP
Catalog No. 5000214812
Encompass_Preferred
Rp-cAMPS sodium salt is a cyclic AMP analog that acts as a competitive antagonist of cAMP-dependent protein kinase A (PKA) isoforms I and II. It is resistant to hydrolysis by phosphodiesterases and is supplied as the sodium salt for biochemical and cell-signaling research applications.
  • Competitive antagonist of PKA I and II with reported Kis in the low micromolar range.
  • Resistant to hydrolysis by phosphodiesterases, improving stability in assays.
  • High reported purity (99.69%).
  • Available as solids in multiple sizes and as a 10 mM solution in DMSO.
  • Suitable for biochemical assays and cellular signaling studies.
Catalog No. 50-002-14812 Supplier Medchemexpress LLC Supplier No. HY100530D10MG
Rp-cAMPS sodium salt is a cyclic AMP analog that acts as a competitive antagonist of cAMP-dependent protein kinase A (PKA) isoforms I and II. It is resistant to hydrolysis by phosphodiesterases and is supplied as the sodium salt for biochemical and cell-signaling research applications.
  • Competitive antagonist of PKA I and II with reported Kis in the low micromolar range.
  • Resistant to hydrolysis by phosphodiesterases, improving stability in assays.
  • High reported purity (99.69%).
  • Available as solids in multiple sizes and as a 10 mM solution in DMSO.
  • Suitable for biochemical assays and cellular signaling studies.