Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rp-cAMPS sodium salt | 142439-94-9 | 99.7% | 367.25 | C10H11N5NaO5PS | 5 MG
SDP

Catalog No. 5000214602
Encompass_Preferred

Rp-cAMPS sodium salt is a cAMP analog that competitively antagonizes cAMP-dependent protein kinase A (PKA) I and II and is resistant to hydrolysis by phosphodiesterases, intended for biochemical and cell signaling research applications.

  • High purity (≈99.7%) suitable for biochemical assays.
  • Molecular formula C10H11N5NaO5PS and molecular weight 367.25 g/mol.
  • Resistant to hydrolysis by phosphodiesterases.
  • Reported Ki values ~6.05 μM (PKA I) and ~9.75 μM (PKA II).
  • Solid, white to pink appearance; available as solid or solution (10 mM in DMSO).
  • Storage: -20°C sealed; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-14602 Supplier Medchemexpress LLC Supplier No. HY100530D5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rp-cAMPS sodium salt is a cAMP analog that competitively antagonizes cAMP-dependent protein kinase A (PKA) I and II and is resistant to hydrolysis by phosphodiesterases, intended for biochemical and cell signaling research applications.

  • High purity (≈99.7%) suitable for biochemical assays.
  • Molecular formula C10H11N5NaO5PS and molecular weight 367.25 g/mol.
  • Resistant to hydrolysis by phosphodiesterases.
  • Reported Ki values ~6.05 μM (PKA I) and ~9.75 μM (PKA II).
  • Solid, white to pink appearance; available as solid or solution (10 mM in DMSO).
  • Storage: -20°C sealed; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.