Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rp-cAMPS sodium salt | 142439-94-9 | 99.7% | 367.25 | C10H11N5NaO5PS | 50 MG
SDP

Catalog No. 5000214778
Encompass_Preferred

Rp-cAMPS sodium salt is a cAMP analog and competitive antagonist of cAMP-dependent protein kinase A (PKA I/II), used to inhibit PKA signaling in biochemical and cellular assays.

  • Acts as a cAMP analog and competitive PKA antagonist.
  • High purity: 99.69%.
  • Molecular formula C10H11N5NaO5PS; molecular weight 367.25.
  • White to pink solid; soluble in water and DMSO.
  • Available in small research quantities including 50 mg.
  • Store sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-14778 Supplier Medchemexpress LLC Supplier No. HY100530D50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rp-cAMPS sodium salt is a cAMP analog and competitive antagonist of cAMP-dependent protein kinase A (PKA I/II), used to inhibit PKA signaling in biochemical and cellular assays.

  • Acts as a cAMP analog and competitive PKA antagonist.
  • High purity: 99.69%.
  • Molecular formula C10H11N5NaO5PS; molecular weight 367.25.
  • White to pink solid; soluble in water and DMSO.
  • Available in small research quantities including 50 mg.
  • Store sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.