Learn More
Medchemexpress LLC Rp-cAMPS sodium salt | 142439-94-9 | 99.7% | 367.25 g/mol | C10H11N5NaO5PS | 1 ML
SDP
Catalog No. 5000214150
Encompass_Preferred
Rp-cAMPS sodium salt is a cyclic AMP analog that competitively antagonizes cAMP-dependent protein kinase A (PKA) isoforms I and II. It is resistant to phosphodiesterase hydrolysis and is supplied as a solid or as a 10 mM solution in DMSO for biochemical and cell signaling studies.
Catalog No. 50-002-14150 Supplier Medchemexpress LLC Supplier No. HY100530D10MM 1ML
Rp-cAMPS sodium salt is a cyclic AMP analog that competitively antagonizes cAMP-dependent protein kinase A (PKA) isoforms I and II. It is resistant to phosphodiesterase hydrolysis and is supplied as a solid or as a 10 mM solution in DMSO for biochemical and cell signaling studies.