Learn More
Medchemexpress LLC Rp-cAMPS sodium salt | 142439-94-9 | 99.7% | 367.25 g/mol | C10H11N5NaO5PS | 100 MG
SDP
Catalog No. 5000214654
Encompass_Preferred
Rp-cAMPS sodium salt is a cyclic AMP analogue that acts as a competitive antagonist of cAMP-dependent protein kinase A (PKA) isoforms I and II. It is resistant to phosphodiesterase hydrolysis and supplied for research use in both solid and solution formats, suitable for biochemical and cell-based assays.
  • Potent competitive antagonist of PKA I and II.
  • Resistant to phosphodiesterase hydrolysis.
  • High purity (≈99.7%) for reliable results.
  • Soluble in water and DMSO with sonication and warming as needed.
  • Available in multiple solid pack sizes and prepared solutions.
  • Stable as a solid at -20°C; solutions stored frozen for limited time.
Catalog No. 50-002-14654 Supplier Medchemexpress LLC Supplier No. HY100530D100MG
Rp-cAMPS sodium salt is a cyclic AMP analogue that acts as a competitive antagonist of cAMP-dependent protein kinase A (PKA) isoforms I and II. It is resistant to phosphodiesterase hydrolysis and supplied for research use in both solid and solution formats, suitable for biochemical and cell-based assays.
  • Potent competitive antagonist of PKA I and II.
  • Resistant to phosphodiesterase hydrolysis.
  • High purity (≈99.7%) for reliable results.
  • Soluble in water and DMSO with sonication and warming as needed.
  • Available in multiple solid pack sizes and prepared solutions.
  • Stable as a solid at -20°C; solutions stored frozen for limited time.