Learn More
Medchemexpress LLC Rp-cAMPS sodium salt | 142439-94-9 | 99.7% | 367.25 g/mol | C10H11N5NaO5PS | 25 MG
SDP
Catalog No. 5000215036
Encompass_Preferred
Rp-cAMPS sodium salt is a stereospecific, non-hydrolyzable cyclic AMP analog that competitively antagonizes cAMP-dependent protein kinase A (PKA) isoforms I and II. It is used as a biochemical tool to probe cAMP/PKA signaling pathways in enzyme inhibition and cell signaling research.
  • Resistant to hydrolysis by phosphodiesterases.
  • Competitive antagonist of cAMP-dependent PKA I and II (Ki ≈ 12.5 μM and 4.5 μM).
  • High reported purity (≈99.7%).
  • Available as small solid quantities and as a DMSO solution.
  • Stable when stored sealed at -20°C; in solution, store at -80°C up to 6 months.
  • Suitable for biochemical assays and cell signaling studies.
Catalog No. 50-002-15036 Supplier Medchemexpress LLC Supplier No. HY100530D25MG
Rp-cAMPS sodium salt is a stereospecific, non-hydrolyzable cyclic AMP analog that competitively antagonizes cAMP-dependent protein kinase A (PKA) isoforms I and II. It is used as a biochemical tool to probe cAMP/PKA signaling pathways in enzyme inhibition and cell signaling research.
  • Resistant to hydrolysis by phosphodiesterases.
  • Competitive antagonist of cAMP-dependent PKA I and II (Ki ≈ 12.5 μM and 4.5 μM).
  • High reported purity (≈99.7%).
  • Available as small solid quantities and as a DMSO solution.
  • Stable when stored sealed at -20°C; in solution, store at -80°C up to 6 months.
  • Suitable for biochemical assays and cell signaling studies.