Learn More
Medchemexpress LLC HY-100530 1mg Medchemexpress, Rp-cAMPS (triethylammonium salt) CAS:151837-09-1 Purity:>98%
SDP
Catalog No. 501967740
Encompass_Preferred
Medchemexpress, HY-100530 1mg Rp-cAMPS (triethylammonium salt) CAS:151837-09-1 Rp-cAMPS triethylammonium salt is an analog of cAMP which acts as a potent, competitive and cell-permeable antagonist of cAMP-induced activation of cAMP-dependent PKA I and II (Kis of 6.05 µM and 9.75 µM, respectively). Rp-cAMPS triethylammonium salt is resistant to hydrolysis by phosphodiesterases[3][4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7740 Supplier Medchemexpress LLC Supplier No. HY1005301MG
Add to Cart
Add to Cart
Medchemexpress, HY-100530 1mg Rp-cAMPS (triethylammonium salt) CAS:151837-09-1 Rp-cAMPS triethylammonium salt is an analog of cAMP which acts as a potent, competitive and cell-permeable antagonist of cAMP-induced activation of cAMP-dependent PKA I and II (Kis of 6.05 µM and 9.75 µM, respectively). Rp-cAMPS triethylammonium salt is resistant to hydrolysis by phosphodiesterases[3][4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.