Learn More
Enzo Life Sciences Guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer . sodium salt (1mg). CAS: 86562-09-6
SDP
Supplier:  Enzo Life Sciences ALX480072M001
Encompass_Preferred
Inhibitor of cGMP-dependent protein kinase (PKG) 1α (cGMP antagonist). Resistant against mammalian cyclic nucleotid-dependent phosphodiesterases. No metabolic side effects. Low membrane permeability. Alternative name: Rp-cGMPS . Na ; cGMPS . Na, Rp-Isomer, cGMPS . Na, Rp-Isomer. Purity: ≥98% (HPLC). Solubility: Soluble in water (125mM), DMSO (2mM), DMF (1mM), methanol (7mM) or PBS, pH 7.4 (125mM). Long Term Storage: +4°C.
Catalog No. 50-200-9316