Learn More
Cayman Chemical Rp-Cyclic AMPS triethylammon
SDP
Catalog No. 502965126
Encompass_Preferred
A non-hydrolyzable phosphorothioate analog of cAMP that competitively inhibits cAMP-dependent protein kinases (PKA) I and II (IC50s = 12.5 and 4.5 µM, respectively); broadly used in research involving cAMP-dependent signaling in vitro and in vivo
Catalog No. 50-296-5126 Supplier Cayman Chemical Supplier No. 16985500 UG
A non-hydrolyzable phosphorothioate analog of cAMP that competitively inhibits cAMP-dependent protein kinases (PKA) I and II (IC50s = 12.5 and 4.5 µM, respectively); broadly used in research involving cAMP-dependent signaling in vitro and in vivo