Learn More
Waters Corp Waters UPLC Method Development Kit, RP and HILIC, 3.0 x 50 mm, 1.7 μm/1.8 μm
Catalog No. 50791076 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This UPLC RP and HILIC Method Development Kit provides a comprehensive selection of column chemistries for efficient method development.
  • Contains four columns per pack.
  • Includes BEH C18, BEH Shield RP18, BEH Amide, and HSS C18 SB chemistries.
  • Features particle sizes of 1.7 μm for BEH columns and 1.8 μm for HSS columns.
  • Columns measure 3.0 x 50 mm.
Catalog No. 50-791-076 Supplier Waters Corp Supplier No. 176001961
Add to Cart
Add to Cart
Request a Quote
This UPLC RP and HILIC Method Development Kit provides a comprehensive selection of column chemistries for efficient method development.
  • Contains four columns per pack.
  • Includes BEH C18, BEH Shield RP18, BEH Amide, and HSS C18 SB chemistries.
  • Features particle sizes of 1.7 μm for BEH columns and 1.8 μm for HSS columns.
  • Columns measure 3.0 x 50 mm.