Learn More
Bon Opus Biosciences Recombinant Human Proline/serine-rich coiled-coil protein 1/PSRC1(C-His) - Source: E. coli
SDP
Supplier:  Bon Opus Biosciences BP277750UG
Encompass_Preferred
Recombinant Human Proline/serine-rich coiled-coil protein 1/PSRC1 is produced by E.coli expression system and the target gene encoding E2-T215 is expressed with a His tag at the C-terminus. Accession#Q6PGN9. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733763