Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rp,R'p)-(R)-Mandyphos | 210842-74-3 | 92.6% | C52H50FeN2P2 | 100 MG
SDP

Catalog No. 5000432868
Encompass_Preferred

(Rp,R'p)-(R)-Mandyphos is a drug intermediate used for the synthesis of various active compounds. It appears as a light yellow to yellow solid.

  • For research use only.
  • Light yellow to yellow solid appearance.
  • Drug intermediate for synthesis of active compounds.
  • Purity of 92.6%.
  • Chemical formula C52H50FeN2P2.
  • Molecular weight 820.76.
  • Store at 4°C, protected from light, and under nitrogen.
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.

Catalog No. 50-004-32868 Supplier Medchemexpress LLC Supplier No. HY79503100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rp,R'p)-(R)-Mandyphos is a drug intermediate used for the synthesis of various active compounds. It appears as a light yellow to yellow solid.

  • For research use only.
  • Light yellow to yellow solid appearance.
  • Drug intermediate for synthesis of active compounds.
  • Purity of 92.6%.
  • Chemical formula C52H50FeN2P2.
  • Molecular weight 820.76.
  • Store at 4°C, protected from light, and under nitrogen.
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.