Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rp,R'p)-(R)-Mandyphos | 210842-74-3 | 92.6% | C52H50FeN2P2 | 250 MG
SDP

Catalog No. 5000358143
Encompass_Preferred

(Rp,R'p)-(R)-Mandyphos is a drug intermediate for the synthesis of various active compounds. This product is for research use only.

  • Drug intermediate for the synthesis of various active compounds
  • Purity: 92.59%
  • Appearance: Solid
  • Color: Light yellow to yellow

Catalog No. 50-003-58143 Supplier Medchemexpress LLC Supplier No. 250MGHY79503
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rp,R'p)-(R)-Mandyphos is a drug intermediate for the synthesis of various active compounds. This product is for research use only.

  • Drug intermediate for the synthesis of various active compounds
  • Purity: 92.59%
  • Appearance: Solid
  • Color: Light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.