Learn More
Bon Opus Biosciences Recombinant Human TNFAIP8-like protein 2(C-His) - Source: E. coli
SDP
Catalog No. NC2733786
Encompass_Preferred
Recombinant Human TNFAIP8-like protein 2 is produced by E.coli expression system and the target gene encoding E2-L184 is expressed with a His tag at the C-terminus. Accession#Q6P589. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733786 Supplier Bon Opus Biosciences Supplier No. BP307850UG
Recombinant Human TNFAIP8-like protein 2 is produced by E.coli expression system and the target gene encoding E2-L184 is expressed with a His tag at the C-terminus. Accession#Q6P589. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4