Learn More
Medchemexpress LLC RP101988 | 2990018-57-8 | 98.8% | 418.45 | 100 MG
SDP
Catalog No. 5000351422
Encompass_Preferred
RP101988 is the major active metabolite of Ozanimod. It is a selective, potent S1PR1 (sphingosine-1-phosphate receptor 1) agonist.
  • Selective and potent S1PR1 agonist
  • EC50s of 0.19 nM for S1PR1 and 32.8 nM for S1PR5
  • Substrate of P-gp and BCRP drug transporters
  • For research use only
Catalog No. 50-003-51422 Supplier Medchemexpress LLC Supplier No. HY136578100MG
RP101988 is the major active metabolite of Ozanimod. It is a selective, potent S1PR1 (sphingosine-1-phosphate receptor 1) agonist.
  • Selective and potent S1PR1 agonist
  • EC50s of 0.19 nM for S1PR1 and 32.8 nM for S1PR5
  • Substrate of P-gp and BCRP drug transporters
  • For research use only