Learn More
Medchemexpress LLC RP101988 | 2990018-57-8 | 98.8% | 418.45 | 50 MG
SDP
Catalog No. 5000351756
Encompass_Preferred
RP101988 is the major active metabolite of Ozanimod and functions as a selective and potent S1PR1 (sphingosine-1-phosphate receptor 1) agonist. It exhibits EC50s of 0.19 nM for S1PR1 and 32.8 nM for S1PR5. It is intended for research use only.
  • Selective and potent S1PR1 agonist
  • EC50s of 0.19 nM for S1PR1 and 32.8 nM for S1PR5
  • Substrate of P-gp and BCRP drug transporters
  • Off-white to light yellow solid appearance
Catalog No. 50-003-51756 Supplier Medchemexpress LLC Supplier No. HY13657850MG
RP101988 is the major active metabolite of Ozanimod and functions as a selective and potent S1PR1 (sphingosine-1-phosphate receptor 1) agonist. It exhibits EC50s of 0.19 nM for S1PR1 and 32.8 nM for S1PR5. It is intended for research use only.
  • Selective and potent S1PR1 agonist
  • EC50s of 0.19 nM for S1PR1 and 32.8 nM for S1PR5
  • Substrate of P-gp and BCRP drug transporters
  • Off-white to light yellow solid appearance